Order Entry
Norway
ContactUsLinkComponent
Anti-Cathepsin D (Ab-1) (BC011) Mouse Monoclonal Antibody [clone: BC011]
Anti-Cathepsin D (Ab-1) (BC011) Mouse Monoclonal Antibody [clone: BC011]
Catalog # IM03-100
Anti-Cathepsin D (Ab-1) (BC011) Mouse Monoclonal Antibody [clone: BC011]
Catalog # IM03-100

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Cathepsin D
  • Antigen symbol:
    CTSD
  • Clonality:
    Monoclonal
  • Clone:
    BC011
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: Cathepsin D (Ab-1) (BC011)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human