Anti-CADM1 Goat polyclonal antibody unconjugated
Catalog # EVBIEB11250
Leverandør: Everest Biotech
Spesifikasjoner
- Antibody type:Primary
- Antigen name:Cell adhesion molecule 1
- Antigen symbol:CADM1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Spesifikasjoner
Om dette produktet
Type: Primary
Antigen: CADM1 (cell adhesion molecule 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: