Anti-ADAM12 Goat Polyclonal Antibody
Katalog # EVBIEB11060
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:ADAM metallopeptidase domain 12
- Antigen symbol:ADAM12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Pig,Rat,Mouse
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: ADAM12 (ADAM metallopeptidase domain 12)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Pig, Rat