Order Entry
Norway
ContactUsLinkComponent
Anti-SLC12A1 / NKCC Goat Polyclonal Antibody
Anti-SLC12A1 / NKCC Goat Polyclonal Antibody
Catalog # EVBIEB09143
Leverandør:  Everest Biotech
undefined
Anti-SLC12A1 / NKCC Goat Polyclonal Antibody
Catalog # EVBIEB09143
Leverandør:  Everest Biotech

Spesifikasjoner

  • Antibody type:
    Primary
  • Antigen symbol:
    Solute carrier family 12, member 1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Epitope:
    internal
  • Cross adsorption:
    none
  • Antigen synonyms:
    NKCC
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

Spesifikasjoner

Om dette produktet

Type: Primary
Antigen: SLC12A1 / NKCC
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Human