Anti-DPM3 Goat Polyclonal Antibody
Catalog # EVBIEB08946
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:Dolichyl-phosphate mannosyltransferase polypeptide 3
- Antigen symbol:DPM3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Western blot:Yes
- Epitope:internal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: DPM3 (dolichyl-phosphate mannosyltransferase polypeptide 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: