Anti-ABCC5 Goat Polyclonal Antibody
Catalog # EVBIEB08699
Supplier: Everest Biotech
Specifications
- Antibody type:Primary
- Antigen name:ATP Binding Cassette, sub-family C, member 5
- Antigen symbol:ABCC5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: ABCC5 (ATP-binding cassette, sub-family C (CFTR/MRP), member 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Goat
Isotype: IgG
Reactivity: