Order Entry
Norway
ContactUsLinkComponent
Anti-EHD2 Goat polyclonal antibody unconjugated
Anti-EHD2 Goat polyclonal antibody unconjugated
  EVBIEB06493
undefined
Anti-EHD2 Goat polyclonal antibody unconjugated
  EVBIEB06493

 

  • Antibody type:
    Primary
  • Antigen name:
    EH-domain containing 2
  • Antigen symbol:
    EHD2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Epitope:
    C-terminal
  • Cross adsorption:
    none
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 µg
  • Pk:
    0,1 mg

 

 

Type: Primary
Antigen: EHD2 (EH-domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Goat
Isotype: IgG
Reactivity: