Anti-TNFRSF11A Mouse Monoclonal Antibody [clone: 64C1385.1]
Catalog # NOVUNBP2-24702
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:tumor necrosis factor receptor superfamily, member 11a, NFKB activator
- Antigen symbol:TNFRSF11A
- Clonality:Monoclonal
- Clone:64C1385.1
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 lambda
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:8792
- Antigen synonyms:ODFR|RANK|OFE|OSTS|FEO|LOH18CR1|OPTB7|CD265|PDB2|TRANCER
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: TNFRSF11A (tumor necrosis factor receptor superfamily, member 11a, NFKB activator)
Clonality: monoclonal
Clone: 64C1385.1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Lambda
Reactivity: Human, Mouse