Anti-SLC22A1 Rabbit Polyclonal Antibody
Catalog # NOVUNBP2-24677
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:POU class 2 homeobox 1
- Antigen symbol:POU2F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:5451
- Antigen synonyms:OCT1|OTF1|oct-1B
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: SLC22A1 (solute carrier family 22 (organic cation transporter), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: