Anti-ZBTB2 Rabbit Polyclonal Antibody
Katalog # NOVUNBP2-24660
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:zinc finger and BTB domain containing 2
- Antigen symbol:ZBTB2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:57621
- Antigen synonyms:ZNF437
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: ZBTB2 (zinc finger and BTB domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: