Anti-ROR gamma Rabbit Polyclonal Antibody
NOVUNBP2-24503
- Antibody type:Primary
- Antigen name:RAR-related orphan receptor C
- Antigen symbol:RORC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Gene ID:6097
- Antigen synonyms:RZRG|NR1F3|RORG|RZR-GAMMA|TOR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: ROR gamma
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: