Anti-ROR gamma Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: 4G419]
NOVUNBP2-24451
- Antibody type:Primary
- Antigen name:RAR-related orphan receptor C
- Antigen symbol:RORC
- Clonality:Monoclonal
- Clone:4G419
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Gene ID:6097
- Antigen synonyms:RZRG|NR1F3|RORG|RZR-GAMMA|TOR
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 Tests
- Pk:100 Tests
Type: Primary
Antigen: ROR gamma
Clonality: monoclonal
Clone: 4G419
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: