Order Entry
Norway
ContactUsLinkComponent
Anti-GAPDH Mouse monoclonal antibody unconjugated [clone: 1A10]
Anti-GAPDH Mouse monoclonal antibody unconjugated [clone: 1A10]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-GAPDH Mouse monoclonal antibody unconjugated [clone: 1A10]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-47339
  • NOVUNBP1-47339SS
  • Antigen symbol:
  • GAPDH
  • GAPDH
  • Antigen name:
  • Glyceraldehyde-3-phosphate dehydrogenase
  • Glyceraldehyde-3-phosphate dehydrogenase
  • Antigen synonyms:
  • GAPDH|G3PD|CDABP0047|GAPD|Glyceraldehyde-3-phosphate dehydrogenase|Peptidyl-cysteine S-nitrosylase GAPDH|HEL-S-162eP|OK/SW-cl.12
  • G3PD|GAPD|HEL-S-162eP
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 1A10
  • 1A10
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2597
  • 2597
  • Isotype:
  • IgG1
  • IgG1
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information