Anti-Ferroportin 1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
NOVUNBP1-21502PE
- Antibody type:Primary
- Antigen name:solute carrier family 40 (iron-regulated transporter), member 1
- Antigen symbol:Solute carrier family 40, member 1
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:30061
- Antigen synonyms:HFE4|SLC11A3|IREG1|MST079|MSTP079|MTP1|FPN1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 μL
- Pk:0,1 mL
Type: Primary
Antigen: Ferroportin 1
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse