Order Entry
Norway
ContactUsLinkComponent
 
Anti-HPa1 Mouse Monoclonal Antibody [clone: DHIC2-2C12]
 
undefined
Anti-HPa1 Mouse Monoclonal Antibody [clone: DHIC2-2C12]

 

  • Catalog No:
  • NOVUNBP1-18949SS
  • NOVUNBP1-18949
  • Antigen symbol:
  • HPa1
  • HPa1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • DHIC2-2C12
  • DHIC2-2C12
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgM
  • IgM
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice