Order Entry
Norway
ContactUsLinkComponent
Anti-HPi4 Mouse Monoclonal Antibody [clone: HIC1-5F10]
Anti-HPi4 Mouse Monoclonal Antibody [clone: HIC1-5F10]
Supplier:  Novus Biologicals
Certificates
undefined
Anti-HPi4 Mouse Monoclonal Antibody [clone: HIC1-5F10]
Supplier:  Novus Biologicals

Specifications

  • Catalog No:
  • NOVUNBP1-18948
  • NOVUNBP1-18948SS
  • Antigen symbol:
  • HPI4
  • HPI4
  • Antigen name:
  • Heterochromatin Protein 4
  • Heterochromatin Protein 4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • HIC1-5F10
  • HIC1-5F10
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Isotype:
  • IgG1
  • IgG1
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information