Anti-CCL2 Rabbit Polyclonal Antibody (DyLight® 488)
Catalog # NOVUNBP1-07035G
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen name:Monocyte Chemoattractant Protein 1
- Antigen symbol:MCP1
- Clonality:Polyclonal
- Conjugation:DyLight® 488
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Gene ID:6347
- Antigen synonyms:monocyte chemoattractant protein-1|small inducible cytokine A2 (monocyte chemotactic protein 1|small inducible cytokine subfamily A (Cys-Cys)|monocyte secretory protein JE|monocyte chemotactic and activating factor|member 2|monocyte chemotactic protein 1|homologous to mouse Sig-je)|small-inducible cytokine A2|monocyte chemoattractant protein 1|C-C motif chemokine 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Specifications
About this item
Type: Primary
Antigen: CCL2 (chemokine (C-C motif) ligand 2)
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Rat