Anti-DYKDDDDK Tag Rat Monoclonal Antibody [clone: L5]
NOVUNBP1-06712
- Antibody type:Tag
- Antigen name:DYKDDDDK tag
- Antigen symbol:DYKDDDDK
- Clonality:Monoclonal
- Clone:L5
- Conjugation:Unconjugated
- Host:Rat
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Multiple
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:500 µL
- Pk:0,5 mL
The DYKDDDDK Epitope Tag Antibody (L5) from Novus Biologicals is a rat monoclonal antibody to DYKDDDDK Epitope Tag. This antibody reacts with human, drosophila, guinea pig, non-species specific. The DYKDDDDK Epitope Tag Antibody (L5) has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen.
There are 2 scientific publications using this antibody.
Type: Primary
Antigen: DYKDDDDK
Clonality: Monoclonal
Clone: L5
Conjugation: Unconjugated
Epitope: DDDDK
Host: Rat
Isotype: IgG2a
Reactivity: Human, Drosophila, Guinea Pig, Non-species specific