Anti-BECN2 Rabbit Polyclonal Antibody
The Beclin 2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Beclin 2. This antibody reacts with human. The Beclin 2 Antibody has been validated for the following applications: Western Blot, Simple Western, Immunohistochemistry-Paraffin.
Simple Western,
Type: Primary
Antigen: Beclin 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Catalog No:
- NOVUNB110-60984
- NOVUNB110-60984SS
- Antigen symbol:
- BECN2
- BECN2
- Antigen name:
- Beclin 2
- Beclin 2
- Antigen synonyms:
- BECN1P1|BECN1L1
- BECN1P1|BECN1L1
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- 441925
- 441925
- Western blot:
- Yes
- Yes
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice