Order Entry
Norway
ContactUsLinkComponent
 
Anti-BAP1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1G8]
  NOVUNB110-60521PE
Anti-BAP1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: 1G8]
  NOVUNB110-60521PE

 

  • Antibody type:
    Primary
  • Antigen name:
    BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase)
  • Antigen symbol:
    BAP1
  • Clonality:
    Monoclonal
  • Clone:
    1G8
  • Conjugation:
    PE (Phycoerythrin)
  • Host:
    Mouse
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    8314
  • Antigen synonyms:
    HUCEP-13|hucep-6|UCHL2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pk:
    100 Tests

 

 

Type: Primary
Antigen: BAP1 (BRCA1 associated protein-1 (ubiquitin carboxy-terminal hydrolase))
Clonality: monoclonal
Clone: 1G8
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human