Anti-BRCA1 Mouse Monoclonal Antibody (HRP (Horseradish Peroxidase)) [clone: RAY]
NOVUNB100-598H
- Antibody type:Primary
- Antigen name:Breast Cancer type 1 susceptibility protein
- Antigen symbol:BRCA1
- Clonality:Monoclonal
- Clone:RAY
- Conjugation:HRP (Horseradish Peroxidase)
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Gene ID:672
- Antigen synonyms:BROVCA1|BRCA1/BRCA2-containing complex|subunit 1|PPP1R53|breast and ovarian cancer sususceptibility protein 1|RING finger protein 53|IRIS|protein phosphatase 1|breast cancer type 1 susceptibility protein|RNF53|PNCA4|regulatory subunit 53|breast and ovarian cancer susceptibility protein 1|PSCP|BRCAI|BRCC1
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µL
- Pk:0,1 mL
Type: Primary
Antigen: BRCA1 (breast cancer 1, early onset)
Clonality: monoclonal
Clone: RAY
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human