Anti-Transferrin Rabbit Polyclonal Antibody
BSBTRP1022
- Antibody type:Primary
- Antigen name:Transferrin
- Antigen symbol:TF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:PRO2086|Beta-1 metal-binding globulin|PRO1557|TFQTL1|Serotransferrin|Siderophilin
- Amino acid number:20 - 698
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:Oryza sativa-derived human Transferrin recombinant protein (Position: V20-P698)
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Serotransferrin(TF) detection. Tested with WB, IHC-P, ELISA in Human.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Transferrin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human