Anti-PLK2 Rabbit Polyclonal Antibody
Katalog # BSBTPA2131
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Polo-like kinase 2
- Antigen symbol:PLK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:PLK-2|Serine/threonine-protein kinase PLK2|Serum-inducible kinase|PLK2|Serine/threonine-protein kinase SNK|Polo-like kinase 1
- Amino acid number:666 - 685
- Sequence:SSELKNRMEYALNMLLQRCN
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human PLK2 (666-685aa SSELKNRMEYALNMLLQRCN), different from the related mouse sequence by one amino acid, and from the related rat sequence by two amino acids
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Serine/threonine-protein kinase PLK2(PLK2) detection. Tested with WB, IHC-P, ICC in Human, Mouse, Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: PLK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat