Order Entry
Norway
ContactUsLinkComponent
Anti-PCSK9 Rabbit Polyclonal Antibody
Anti-PCSK9 Rabbit Polyclonal Antibody
Catalog # BSBTPA2130
Supplier:  Boster Bio
undefined
Anti-PCSK9 Rabbit Polyclonal Antibody
Catalog # BSBTPA2130
Supplier:  Boster Bio

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Proprotein convertase subtilisin/kexin type 9
  • Antigen symbol:
    PCSK9
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Form:
    Lyophilized
  • Antigen synonyms:
    PCSK9|Neural apoptosis-regulated convertase 1|NARC-1|NARC1|Proprotein convertase subtilisin/kexin type 9|Subtilisin/kexin-like protease PC9|Proprotein convertase PC9|PSEC0052
  • Amino acid number:
    671 - 687
  • Sequence:
    AVTAVAICCRSRHLAQA
  • Storage temperature:
    –20 °C
  • Shipping temperature:
    Ice
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human PCSK9 (671-687aa AVTAVAICCRSRHLAQA)
  • Purification:
    Purified by immunogen affinity purification
  • Size:
    100 µg
  • Pk:
    0,1 mg

Specifications

About this item

Rabbit IgG polyclonal antibody for Proprotein convertase subtilisin/kexin type 9(PCSK9) detection. Tested with WB, IHC-P in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: PCSK9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human