Anti-Cdk7 Rabbit Polyclonal Antibody
Katalog # BSBTPA2126-2
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Cyclin-dependent kinase 7
- Antigen symbol:CDK7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:CAK1|MO15|Cell division protein kinase 7|CDK7|CAK|TFIIH basal transcription factor complex kinase subunit|39 kDa protein kinase|P39 Mo15|STK1|CDK-activating kinase
- Amino acid number:329 - 346
- Sequence:RKRAEALEQGILPKKLIF
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of mouse Cdk7 (329-346aa RKRAEALEQGILPKKLIF), identical to the related rat sequence, and different from the related human sequence by two amino acids
- Purification:Purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Cyclin-dependent kinase 7(CDK7) detection. Tested with WB in Human, Rat, Mouse.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Cdk7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse