Anti-Tec Rabbit Polyclonal Antibody
BSBTPA2000
- Antibody type:Primary
- Antigen name:Tec protein tyrosine kinase
- Antigen symbol:TEC
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:PSCTK4|MGC126762|MGC126760|TEC
- Amino acid number:612 - 631
- Sequence:FEDLLRTIDELVECEETFGR
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human Tec (612-631aa FEDLLRTIDELVECEETFGR), identical to the related rat sequence, and different from the related mouse sequence by one amino acid.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Rabbit IgG polyclonal antibody for Tyrosine-protein kinase Tec (TEC) detection. Tested with WB, IHC-P in Human; Mouse; Rat.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Tec
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat