Anti-Resistin Rabbit Polyclonal Antibody
Catalog # BSBTPA1473
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Resistin
- Antigen symbol:RETN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilized
- Antigen synonyms:Cysteine-rich secreted protein FIZZ3|Adipose tissue-specific secretory factor|C/EBP-epsilon-regulated myeloid-specific secreted cysteine-|RSN|ADSF
- Amino acid number:93 - 114
- Sequence:CHCQCARIDWTAARCCKLQVAS
- Storage temperature:–20 °C
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of mouse Resistin (93-114aa CHCQCARIDWTAARCCKLQVAS).
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:0,1 mg
Specifications
About this item
Rabbit IgG polyclonal antibody for Resistin(RETN) detection. Tested with WB, IHC-P in Mouse.
Reconstitute with distilled water.
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: Resistin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Mouse