Order Entry
Norway
ContactUsLinkComponent
Anti-Thymidylate Synthase Rabbit Monoclonal Antibody [clone: SP112]
Anti-Thymidylate Synthase Rabbit Monoclonal Antibody [clone: SP112]
Supplier:  Abcam
Certificates
undefined
Anti-Thymidylate Synthase Rabbit Monoclonal Antibody [clone: SP112]
Supplier:  Abcam

Specifications

  • Catalog No:
  • ABCAAB111153-100
  • ABCAAB111153-1
  • ABCAAB111153-500
  • Antigen symbol:
  • TYMS
  • Antigen name:
  • Thymidylate Synthase
  • Antibody type:
  • Primary
  • Clonality:
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Clone:
  • SP112
  • Host:
  • Rabbit
  • Isotype:
  • IgG
  • ImmunoChemistry:
  • Yes
  • Storage temperature:
  • Cool
  • Shipping temperature:
  • Ice

Specifications