Order Entry
Norway
ContactUsLinkComponent
Anti-ROR gamma Mouse Monoclonal Antibody [clone: 4G419] (FITC (Fluorescein Isothiocyanate))
Anti-ROR gamma Mouse Monoclonal Antibody [clone: 4G419] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCAAB104906-100
Leverandør:  Abcam
undefined
Anti-ROR gamma Mouse Monoclonal Antibody [clone: 4G419] (FITC (Fluorescein Isothiocyanate))
Catalog # ABCAAB104906-100
Leverandør:  Abcam

Spesifikasjoner

  • Antibody type:
    Primary
  • Antigen symbol:
    ROR gamma
  • Clonality:
    Monoclonal
  • Clone:
    4G419
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Seq:
    0790
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    100 Tests
  • Pack type:
    Vial
  • Pk:
    100 Tests

Spesifikasjoner

Om dette produktet

FITC Mouse monoclonal [4G419] to ROR gamma, prediluted.

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ROR gamma
Clonality: monoclonal
Clone: 4G419
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: