Specifications
- Antibody type:Primary
- Antigen name:ADP-ribosylation factor 1
- Antigen symbol:ARF1
- Clonality:Monoclonal
- Clone:ARFS 3F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:100 µg
- Pk:50 µG
Specifications
About this item
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ARF1 (ADP-ribosylation factor 1)
Clonality: monoclonal
Clone: ARFS 3F1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity: Human