Anti-Protein Z Mouse Monoclonal Antibody
BIRBORB95429-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Protein Z
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Format:ascites fluid
- Storage buffer:0,03% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
ELISA: 1:10000; Immunochemistry: 1:200-1:400; Western Blot: 1:500-1:2000
Type: Primary
Antigen: Protein Z
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human