Anti-TLR7 Rabbit Polyclonal Antibody
BIRBORB89973-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Toll-like Receptor 7
- Antigen symbol:TLR7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Western blot:Yes
- Storage buffer:PBS, 0,2% gelatin, 0,05% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:50 µg
- Pk:50 µG
Immunochemistry: 10 ?g/ml; Western Blot: 0.5-2 ?g/ml
Type: Primary
Antigen: TLR7 (toll-like receptor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: