Anti-RCHY1 Mouse Monoclonal Antibody
BIRBORB89427-50
: Biorbyt
- Antibody type:Primary
- Antigen name:Ring finger and CHY zinc finger domain containing 1, E3 ubiquitin protein ligase
- Antigen symbol:RCHY1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Western blot:Yes
- Format:ascites fluid
- Storage buffer:0,03% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
ELISA: 1:10000; Flow Cytometry: 1:200-1:400; Immunofluorescence: 1:200-1:1000; Immunochemistry: 1:200; Western Blot: 1:500-1:2000
Type: Primary
Antigen: RCHY1 (ring finger and CHY zinc finger domain containing 1, E3 ubiquitin protein ligase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: