Order Entry
Norway
ContactUsLinkComponent
Anti-Penicillin Mouse Monoclonal Antibody
Anti-Penicillin Mouse Monoclonal Antibody
Catalog # BIRBORB76773-0.1
Supplier:  Biorbyt
Anti-Penicillin Mouse Monoclonal Antibody
Catalog # BIRBORB76773-0.1
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Penicillin
  • Antigen symbol:
    PCN
  • Clonality:
    Monoclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Storage buffer:
    Purified antibody containing PBS, 0,1% sodium azide
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    Bovine gamma-globulin conjugated benzylpenicillin
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 μL
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: Penicillin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: