Anti-NQO1 Mouse Monoclonal Antibody
Catalog # BIRBORB76743-50
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:NAD(P)H dehydrogenase, quinone 1
- Antigen symbol:NQO1
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Storage buffer:Purified antibody containing PBS, 0,1% sodium azide
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Full length purified recombinant human NQO1
- Purification:purified by Protein G chromatography
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Western Blot: 1:2000
Type: Primary
Antigen: NQO1 (NAD(P)H dehydrogenase, quinone 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human