Anti-NOX1 Rabbit Polyclonal Antibody
Katalog # BIRBORB76252-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:NADPH oxidase 1
- Antigen symbol:NOX1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:lyophilized
- Amino acid number:354 to 374
- Storage buffer:5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
- Sequence:GDWTENLIRAFEQQYS
- Storage temperature:Cool
- Concentration:0,1-0,5 ug/ml,0,5-1 ug/ul
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence in the middle region of human NOX1(354-374aa, HIRAAGDWTENLIRAFEQQYS).
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: NOX1 (NADPH oxidase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human