Anti-BANP Rabbit Polyclonal Antibody
Catalog # BIRBORB75495-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:BTG3 associated nuclear Protein
- Antigen symbol:BANP
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Storage temperature:Cool
- Concentration:1 mg/mL
- Shipping temperature:Ice
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: BANP (BTG3 associated nuclear protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat