Anti-SCN9A Mouse Monoclonal Antibody
Catalog # BIRBORB67404-100
Leverandør: Biorbyt
Spesifikasjoner
- Antibody type:Primary
- Antigen name:sodium channel, voltage-gated, type IX, alpha subunit
- Antigen symbol:SCN9A
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Amino acid number:1751 to 1946
- Storage buffer:50% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Fusion protein amino acids 1751-1946 (C-terminus) of human Nav1.7, accession number Q15858
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Spesifikasjoner
Om dette produktet
Western Blot: 1-10 ?g/ml; Immunochemistry: 0.1-1.0 ?g/ml; Immunofluorescence: 1.0-10 ?g/ml
Type: Primary
Antigen: SCN9A (sodium channel, voltage-gated, type IX, alpha subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat