Anti-SLC9A2 Rabbit Polyclonal Antibody
Catalog # BIRBORB138018-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:solute carrier family 9, subfamily A (NHE2, cation proton antiporter 2), member 2
- Antigen symbol:Solute carrier family 9, member 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:lyophilized
- Storage buffer:5mg BSA, 0,9mg NaCl, 0,2mg Na2HPO4, 0,05mg Thimerosal, 0,05mg NaN3.
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human SLC9A2, different from the related rat and mouse sequences by two amino acids.
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: SLC9A2 (solute carrier family 9, subfamily A (NHE2, cation proton antiporter 2), member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat