Order Entry
Norway
ContactUsLinkComponent
Anti-SERPINA1 Rabbit Polyclonal Antibody
Anti-SERPINA1 Rabbit Polyclonal Antibody
Supplier:  Biorbyt
Certificates
undefined
Anti-SERPINA1 Rabbit Polyclonal Antibody
Supplier:  Biorbyt

Specifications

  • Catalog No:
  • BIRBORB129737-100
  • BIRBORB129737-200
  • Antigen symbol:
  • SERPINA1
  • SERPINA1
  • Antigen name:
  • Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1
  • Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Polyclonal
  • Polyclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Host:
  • Rabbit
  • Rabbit
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • Immunogen:
  • Recombinant Protein
  • Recombinant Protein
  • Purification:
  • purified by affinity chromatography
  • purified by affinity chromatography
  • Concentration:
  • 0,9 mg/ml
  • 0,9 mg/ml
  • Storage temperature:
  • Cool
  • Cool
  • Shipping temperature:
  • Ice
  • Ice

Specifications

Related Information