Anti-NMNAT3 Goat Polyclonal Antibody
BIRBORB125180-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Nicotinamide mononucleotide adenylyltransferase 3
- Antigen symbol:NMNAT3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Goat
- Reactivity:Mouse
- Western blot:Yes
- Storage buffer:Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Molecular weight:28 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:internal region
- Purification:purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Size:100 µg
- Pk:100 µG
ELISA: 1:64000; Western Blot: 0.5-2 ?g/ml
Type: Primary
Antigen: NMNAT3 (nicotinamide nucleotide adenylyltransferase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse