Anti-MAPK8/9/10 Rabbit Polyclonal Antibody
Catalog # BIRBORB106161-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Mitogen-activated protein kinase 8/9/10
- Antigen symbol:MAPK8/9/10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Modification:phospho-Thr183, phospho-Tyr185
- Storage buffer:Phosphate buffer saline, pH. 7.4, 150mM NaCl, 0,02% sodium azide and 50% glycerol.
- Molecular weight:46, 54 kDa
- Storage temperature:Cool
- Concentration:20 ug: 0,2 mg/ml; 50 ug: 0,5 mg/ml; 100 ug: 1 ug/ul; 200 ug: 1 ug/ul
- Shipping temperature:Ice
- Immunogen:A synthesized peptide derived from human JNK1/2/3 around the phosphorylation site of Threonine 183+Tyrosine 185
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Western Blot: 1:500-1:2000
Type: Primary
Antigen: MAPK8/9/10 (mitogen-activated protein kinase 8/9/10)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat