Anti-HIV-1 gp24 Mouse monoclonal antibody
Catalog # BIRBORB44463-0.1
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen symbol:HIV-1 gp24
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Western blot:Yes
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified
- Size:100 μg
- Pk:0,1 mg
Specifications
About this item
Type: Primary
Antigen: HIV-1 gp24
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: