Anti-PIN-Cocktail 2 P504S / p63 / HMW Cytokeratin None Monoclonal Antibody
: Biorbyt
Type: Primary
Antigen: PIN-Cocktail 2 P504S / p63 / HMW Cytokeratin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: None
Isotype: IgG+IgG2a+IgG1
Reactivity: Human
- Catalog No:
- BIRBORB27519-0.5
- BIRBORB27519-1
- Antigen symbol:
- P63
- P63
- Antigen name:
- PIN-Cocktail 2 P504S
- PIN-Cocktail 2 P504S
- Antigen synonyms:
- HMW Cytokeratin
- HMW Cytokeratin
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Reactivity:
- Human
- Human
- Isotype:
- IgG, IgG2a, IgG1
- IgG, IgG2a, IgG1
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- 1. Human recombinant AMACR (P504S) protein, whole sequence (for rabbit monoclonal antibody 13H4); 2. Recombinant human p63 protein aa 1-205 (for mouse monoclonal antibody 4A4)
- 1. Human recombinant AMACR (P504S) protein, whole sequence (for rabbit monoclonal antibody 13H4); 2. Recombinant human p63 protein aa 1-205 (for mouse monoclonal antibody 4A4)
- Format:
- concentrated
- concentrated
- Form:
- liquid
- liquid
- Concentration:
- 0.0694444444
- 0.0694444444
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice