Anti-TPO Mouse Monoclonal Antibody
Catalog # BIRBORB24290-1
Leverandør: Biorbyt
Spesifikasjoner
- Antibody type:Primary
- Antigen name:Thyroid Peroxidase
- Antigen symbol:TPO
- Clonality:Monoclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Storage buffer:PBS, pH 7,4
- Storage temperature:Cool
- Concentration:4,3 mg/ml (Sigma Protein Assay Kit)
- Shipping temperature:Ice
- Immunogen:Human Thyroid Peroxidase
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Spesifikasjoner
Om dette produktet
Type: Primary
Antigen: TPO (thyroid peroxidase)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human