Order Entry
Norway
ContactUsLinkComponent
 
Anti-IgG lambda Sheep Polyclonal Antibody
  BIRBORB22436-1
 :  Biorbyt
Anti-IgG lambda Sheep Polyclonal Antibody
  BIRBORB22436-1
 :  Biorbyt

 

  • Antibody type:
    Secondary
  • Antigen name:
    lambda light chain
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Sheep
  • Isotype:
  • Reactivity:
  • Environmentally Preferable:
  • Format:
    antiserum
  • Form:
    lyophilized
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    A pool of purified Bence Jones lambda proteins isolated from human urine. Freunds complete adjuvant is used in the first step of the immunization procedure.
  • Size:
    1 ml
  • Pk:
    1 mL

 

 

Type: Secondary
Antigen: lambda light chain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: none
Reactivity: