Anti-BF2 Goat Polyclonal Antibody
Supplier: Biorbyt
Certificates
About this item
ELISA: 1:64000; Western Blot: 0.3-1.0 ?g/ml
Type: Primary
Antigen: BF2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Specifications
- Catalog No:
- BIRBORB18407-20
- BIRBORB18407-100
- Antigen symbol:
- BF2
- BF2
- Antibody type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Host:
- Goat
- Goat
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Immunogen:
- C Terminus
- C Terminus
- Molecular weight:
- 48 kDa
- 48 kDa
- Purification:
- purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- purified by ammonium sulphate precipitation followed by antigen affinity chromatography
- Storage buffer:
- Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Supplied at 0,5 mg/ml in Tris saline, 0,02% sodium azide, pH 7,3 with 0,5% BSA
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice