Anti-RUNX2 Rabbit Polyclonal Antibody
Catalog # BIRBORB4788-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:Runt-related Transcription Factor 2
- Antigen symbol:RUNX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:liquid
- Modification:phospho-Ser451
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:67 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human RUNX2 around the phosphorylation site of Ser451.
- Purification:purified by affinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500
Type: Primary
Antigen: RUNX2 (runt-related transcription factor 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat