Anti-PSMA8 Mouse Monoclonal Antibody [clone: 2E3]
ABNOH00143471-M03
: Abnova
- Antibody type:Primary
- Antigen name:Proteasome 20S Alpha 8
- Antigen symbol:PSMA8
- Clonality:Monoclonal
- Clone:2E3
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Cross adsorption:No
- Gene ID:143471
- Antigen synonyms:PSMA7L
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:50 µg
- Pk:100 µg
Type: Primary
Antigen: PSMA8
Clonality: Monoclonal
Clone: 2E3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat