Anti-MED8 Mouse Polyclonal Antibody
ABNOH00112950-B01
: Abnova
- Antistofftype:Primary
- Antigen-navn:Mediator Complex Subunit 8
- Antigensymbol:MED8
- Klonalitet:Polyclonal
- Konjugering:Unconjugated
- Vert:Mouse
- Reaktivitet:Human
- Western blot:Yes
- Kyssabsorpsjon:No
- Gen-ID:112950
- Antigen synonymer:ARC32
- Lagringstemp:Cool
- Frakttemp.:Ice
- Str:50 µl
- Fp:50 µl
Type: Primary
Antigen: MED8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human